Tisk…
Spin filter plates, desalting, 96-well, Zeba™
Dodavatel: Thermo Fisher Scientific
PIER87775EA 21940 CZK
PIER87775 PIER87774
Spin filter plates, desalting, 96-well, Zeba™
Mikrodestičky Destičky filtrační
Zeba™ spin desalting plates are polypropylene devices containing a proprietary high performance size exclusion resin for effective removal of salts and other small molecules.
  • High recovery, low-binding resin maximizes protein recovery
  • No screening of fractions or waiting for protein to emerge by gravity-flow
  • Excellent recovery for protein larger than 40000 Da
These plates contain a size-exclusion chromatography resin that provides excellent protein desalting performance in a centrifuge format for 5 µl to 4 ml samples. This 96-well spin plate format enables more high-throughput processing. Require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 µl) in 5 minutes.