Tisk…

Ethyl-trans-2-amino-4-cyclohexene-1-carboxylate hydrochloride 98%

Dodavatel: Thermo Scientific

ACRO334410010EA 7910 CZK
ACRO334410010
Ethyl-trans-2-amino-4-cyclohexene-1-carboxylate hydrochloride 98%
Ethyl-trans-2-amino-4-cyclohexene-1-carboxylate hydrochloride

Appearance: White to beige to grey Powder

Vzorec: C₉H₁₆ClNO₂
Bod tání: 107…109 °C
Storage Temperature: Ambient
CAS číslo: 142547-16-8

Objednat

Výsledky specifikačního testu

Appearance White to beige to grey Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR