Tisk…

(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 98%

Dodavatel: Thermo Scientific

ACRO125130250EA 2020 CZK
ACRO125130250
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 98%
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride

Appearance: White to yellowish-white Powder or crystals

Vzorec: C₇H₁₅NO₂·HCl
MW: 181,7 g/mol
Bod tání: 145…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012494
CAS číslo: 7517-19-3

Objednat

Výsledky specifikačního testu

Appearance White to yellowish-white Powder or crystals
Infrared spectrum Conforms
Specific optical rotation +18.5° to +21.5° (20°C, 589 nm) (c=4.5, methanol)
Titration with HClO4 ≥97.5 %
Residue after evaporation ≤0.005 %
UV at 200 nm A: ≤0.75
UV 1 cm cell vs HPLC-grade water
Non-volatile matter ≤0.0003 %
Free acid (CH3COOH) ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR