Tisk…

trans-Diethyl caronate 90%

Dodavatel: Thermo Scientific

ACRO374260010EA 7250 CZK
ACRO374260010
trans-Diethyl caronate 90%
trans-Diethyl caronate

Appearance: Clear colorless to light yellow Liquid

Vzorec: C₁₁H₁₈O₄
Storage Temperature: Ambient
CAS číslo: 67945-22-6

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4450 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR