3-Fluorphenylmagnesium bromide 1 M in THF, AcroSeal™
Dodavatel: Thermo Scientific
|
Nebezpečí
|
427821000.EA
6220
CZK
427821000.
3-Fluorphenylmagnesium bromide 1 M in THF, AcroSeal™
3-Fluorphenylmagnesium bromide
Vzorec:
C₆H₄BrFMg Bod vzplanutí: –21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672007 CAS číslo: 17318-03-5 UN: 3399 ADR: 4.3,II |
Výsledky specifikačního testu
Appearance | Yellow to brown Solution |
Titration Grignard | 1 M Exact strength on the certificate of analysis |
Refractive index | 1.4240 to 1.4270 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...