Tisk…
High-Performance Lab Freezers, Forma™
Dodavatel: Thermo Scientific
HERAFFGL1230VEA 166060 CZK
HERAFFGL1230V
High-Performance Lab Freezers, Forma™
Forma™ high-Performance Lab Freezers, designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals and other commonly used laboratory materials.
  • Rugged, long-life cabinets with smooth, scratch-resistant interiors and exteriors that are easy to clean
  • Directed airflow for fast temperature recovery after door openings
  • Solid, self-closing doors with keyed lock
  • Battery backup and remote alarm connections
  • Alarm silence, ringback and automatic reset functions