Pyridine-3-sulphonyl chloride hydrochloride 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
ACRO451480050EA
7650
CZK
ACRO451480050
Pyridine-3-sulphonyl chloride hydrochloride 95%
Pyridine-3-sulphonyl chloride hydrochloride
Vzorec:
C₅H₅Cl₂NO₂S Bod tání: 144…145 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04035552 CAS číslo: 42899-76-3 UN: 3261 ADR: 8,III |
Výsledky specifikačního testu
Appearance | White to yellow Powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % as diethylsulfonamide |
Elemental Analysis | 26.37 to 29.74 % (C) |
Elemental Analysis | 2.21 to 2.50 % (H) |
Elemental Analysis | 6.15 to 6.94 % (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...