Tisk…
Hledali jste: Standardy pro ropný průmysl
164  výsledků nalezeno
SearchResultCount:"164"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: VWR Chemicals
Popis: Fuel testing Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products. This ...
Katalogové číslo: (91688.230)
Dodavatel: VWR Chemicals
Popis: Nominal flash point standard (65 °C), VWR®, Glass bottle
Měrná jednotka: 1 * 250 mL
Katalogové číslo: (99104.230)
Dodavatel: VWR Chemicals
Popis: Cloud Point Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Clo...
Měrná jednotka: 1 * 250 mL
Dodavatel: VWR Chemicals
Popis: Total Base Number (TBN) standards are specifically manufactured for the verification of analytical instruments used to determine base number by potent...
Dodavatel: VWR Chemicals
Popis: Sulphur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulphur content in petroleum produ...
Dodavatel: VWR Chemicals
Popis: Sulphur CRMs are suitable for calibration, verification, or for use in quality control procedures used for analysing sulphur content in petroleum prod...
Dodavatel: VWR Chemicals
Popis: Total Acid Number (TAN) standards are specifically manufactured for the verification of analytical instruments used to determine acid number by potent...
Dodavatel: VWR Chemicals
Popis: Sulphur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulphur content in petroleum produ...
Dodavatel: VWR Chemicals
Popis: Sulphur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulphur content in petroleum produ...
Dodavatel: VWR Chemicals
Popis: Sulphur CRMs are suitable for calibration, verification, or for use in quality control procedures used for analysing sulphur content in petroleum prod...
Dodavatel: VWR Chemicals
Popis: Polysulphide CRMs are suitable for calibration, verification, or for use in quality control procedures used for analysing sulphur content in petroleum...
Dodavatel: VWR Chemicals
Popis: Flash point Certified Reference Materials are for the verification of laboratory flash point instruments that are used in testing petroleum and deriva...
Dodavatel: VWR Chemicals
Popis: Pour Point Certified Reference Materials for the verification of laboratory instruments used in testing petroleum and derivative products for Pour Poi...
Dodavatel: VWR Chemicals
Popis: COC flash point standards are tested and certified in accordance with ASTM D92 'Standard Test Method for Flash and Fire Points by Cleveland Open Cup T...
Dodavatel: VWR Chemicals
Popis: PMCC flash point standards are tested and certified in accordance with ASTM D93 'Standard Test Method for Flash Point by Pensky Martens Closed Cup Tes...
Dodavatel: VWR Chemicals
Popis: Flash point Certified Reference Materials are for the verification of laboratory flash point instruments that are used in testing petroleum and deriva...
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 16 of 164
no targeter for Bottom