HPLC columns, ZORBAX® Bonus-RP
Supplier: Agilent
Certificates
About this item
The ZORBAX® Bonus-RP column has a polar amide group embedded in a long alkyl chain. This novel bonding reduces interactions between basic compounds and the silica support, improving peak shape for the most difficult basic compounds. Peak shape and column lifetime are further improved by triple endcapping. In addition, diisopropyl side groups provide steric protection against acid hydrolysis for good lifetime and low pH. The Bonus-RP column provides an alternate selectivity to C18 and C8 alkyl bonded phases.
- Excellent peak shape for challenging basic compounds at low and mid pH
- Unique reversed-phase selectivity
- Novel bonding technology with embedded polar group and steric protection
- Usable in 100% aqueous mobile phases
Specifications
- Catalog No:
- HEWL828668-301
- HEWL827668-301
- HEWL827768-901
- HEWL830668-901
- HEWL883668-901K
- HEWL827668-901K
- HEWL827668-301K
- HEWL884950-577
- HEWL864668-901K
- HEWL828668-901
- HEWL828668-901K
- HEWL828668-301K
- HEWL863668-901K
- HEWL857768-901
- HEWL835668-901K
- HEWL880668-901K
- Carbon load:
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- Endcapped:
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- Triple
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- 180 m²/g
- Temperature limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
Specifications
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL827668-301K827668-301KMethod Validation KitBonus-RP80 Å1,8 µm3,00 mm50 mmLog in for availability1 * 1 KS77 380,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL828668-301K828668-301KMethod Validation KitBonus-RP80 Å1,8 µm3,00 mm100 mmLog in for availability1 * 1 KS88 640,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL828668-901K828668-901KMethod Validation KitBonus-RP80 Å1,8 µm4,00 mm100 mmLog in for availability1 * 1 KS89 520,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL827668-901K827668-901KMethod Validation KitBonus-RP80 Å1,8 µm4,60 mm50 mmLog in for availability1 * 1 KS77 380,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL835668-901K835668-901KMethod Validation KitBonus-RP80 Å3,5 µm4,60 mm50 mmLog in for availability1 * 1 KS62 680,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL864668-901K864668-901KMethod Validation KitBonus-RP80 Å3,5 µm4,60 mm100 mmLog in for availability1 * 1 KS72 190,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL863668-901K863668-901KMethod Validation KitBonus-RP80 Å3,5 µm4,60 mm150 mmLog in for availability1 * 1 KS87 230,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL883668-901K883668-901KMethod Validation KitBonus-RP80 Å5,0 µm4,60 mm150 mmLog in for availability1 * 1 KS76 640,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C - HEWL880668-901K880668-901KMethod Validation KitBonus-RP80 Å5,0 µm4,60 mm250 mmLog in for availability1 * 1 KS86 750,00 KčSpecifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load9,5%EndcappedTriplepH range2 - 9Surface area180 m²/gTemperature limit60 °C