Tisk…
Mop frame, Clino® Ergo EM
Dodavatel: Pfennig
PPSA2520150EA 1080 CZK
PPSA2520150
Mop frame, Clino® Ergo EM
These mop frames are used in hygienically sensitive areas for ergnomically correct cleaning procedures.
  • Autoclavable
  • Completely closed surfaces (hygienic design)
  • High resilience through innovative material
  • Low weight makes it perfect for wall and ceiling cleaning