Voda for LC-MS, Pierce™
Dodavatel: Thermo Fisher Scientific
PIER85189EA
9930
CZK
PIER85189
Voda for LC-MS, Pierce™
Voda
Water with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.
- Purity and quality validated with 30 tests for use in LC-MS applications
- 0.2-micron filtered
- Sealed under nitrogen atmosphere
- High consistency and reliability
Vzorec:
H₂O MW: 18,02 g/mol Bod varu: 100 °C (1013 hPa) Bod tání: 0 °C Hustota: 1 g/cm³ (4 °C) |
MDL Number:
MFCD00011332 CAS číslo: 7732-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...