Tisk…
Labels for TLS 2200®/TLS PC Link™ Printers
Dodavatel: Brady
BRDY011178EA 2990 CZK
BRDY011178 BRDY018519 BRDY018631 BRDY018423 BRDY018544 BRDY018624 BRDY018448 BRDY018526 BRDY018625 BRDY018623 BRDY018680 BRDY018681 BRDY018351 BRDY011181 BRDY011180
Labels for TLS 2200®/TLS PC Link™ Printers
Štítky
Permanent acrylic labels.
  • Continuous tape is not cut to length, it is cut by the printer
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
Labels for the TLS2200® and TLS PC Link™ thermal transfer portable printers. TLS2200® parts can also be printed on the BMP™71 Label printer, provided the roll adapter (M71-ADAPT) is used.