Tisk…
Ion selective electrode, half-cell, ammonium, sensION+ 9663
Dodavatel: Hach
sensION™+
HACHLZW9663970002EA 29970 CZK
HACHLZW9663970002
Ion selective electrode, half-cell, ammonium, sensION+ 9663
sensION+ 9663 is a half-cell (reference not integrated) ammonium ion selective electrode with an epoxy body and non-replaceable, solid-state PVC (polymer) membrane selective to ammonium ions in solution.
  • Utilises a unique solid-state sensor technology that eliminates costly membrane replacements
  • Requires virtually no maintenance: Solid-state sensor design allows for dry storage of the ISE without a shelf life or membrane replacement
  • Provides fast, stable, and accurate response in a variety of sample types
The 9663 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach sensION+ MM374 multimeter. The electrode is ideal for measuring ammonium concentrations in drinking water, wastewater and general water quality applications.
Informace o dodávce: Supplied with basic user manual.