CurveX 3 Standard is a 6-channel thermocouple K type input oven is used to monitoring the progress of the burning process of powder paint kilns.
- Equipped with a large colour touch screen interface for easy operation
- Operation via the menu on the display device
- High-precision temperature logging
- Measurement, analysis and reports are fully user-selectable
- Compatible with ideal finish analysis software
- Aluminium housing with protective sleeve
The data logger is fitted with a large full-color touchscreen for easy menu-driven operation and quick display of measurement results. CurveX 3 standard oven logger kit contains all USB cable, USB charger, USB stick with ideal finish software, probe ID kit, calibration certificate, manual CurveX 3 standard, manual ideal finish analysis, plastic carrying case, CX2005 insulation box, CX2011 heat absorber, just add the desired magnetic or clamp-type probes to make the oven logger KIT complete.
Informace o dodávce: Supplied with USB cable and charger, USB memory stick with Ideal Finish Analysis software, set of identification labels probes, factory calibrated calibration certificate and plastic case.