Tisk…
Hot wire anemometer
Dodavatel: Industrial Physics
SIMELU8050EA 11340 CZK
SIMELU8050
Hot wire anemometer
Hot wire anemometer is ideal for low air velocity and volume measurements.
  • Measures the air velocity in m/s, km/hr, ft/min, MPH and knots
  • Delivers fast and accurate measurements
  • Combination of hot wire and standard thermistor delivers fast and accurate measurements
Informace o dodávce: Supplied with a built-in temperature probe.