Tisk…

Anti-FOXH1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3420-100EA 8430 CZK
BIOV3420-100
Anti-FOXH1 Rabbit Polyclonal Antibody
FAST (Fas-activated serine/threonine) is a novel serine/threonine kinase that is activated as a result of Fas ligation. FAST resides in the cytoplasm as a highly phosphorylated protein and is quickly dephosphorylated and activated in response to stimulated Fas. FAST shows kinase specificity towards both TIA-1 and TIAR, both of which are thought to function as mediators of apoptotic cell death.

Type: Primary
Antigen: FOXH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR