Tisk…

Anti-GZMB Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6683-100EA 7780 CZK
BIOV6683-100 BIOV6683-30T
Anti-GZMB Rabbit Polyclonal Antibody
This enzyme is necessary for target cell lysis in cell-mediated immune responses. It cleaves after Asp. Seems to be linked to an activation cascade of caspases (aspartate-specific cysteine proteases) responsible for apoptosis execution. Cleaves caspase-3, -7, -9 and 10 to give rise to active enzymes mediating apoptosis.

Type: Primary
Antigen: GZMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR