Tisk…

Anti-TNFRSF9 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5370-30TEA 2850 CZK
BIOV5370-30T
Anti-TNFRSF9 Rabbit Polyclonal Antibody
Human 4-1BB Receptor (4-1BBR) is an inhibitor of 4-1BBL and can block 4-1BBL bioactivity. Human 4-1BBR is a 17.7 kDa protein containing 167 amino acid residues.

Type: Primary
Antigen: TNFRSF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR