Tisk…

Anti-FABP5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5505-30TEA 2850 CZK
BIOV5505-30T
Anti-FABP5 Rabbit Polyclonal Antibody
FABPs have been reported to be involved in the uptake and metabolism of fatty acids, maintenance of cellular membrane fatty acids levels, intracellular trafficking, modulation of specific enzymes of lipid metabolic pathways, as well as in the modulation of cell growth and differentiation.

Type: Primary
Antigen: FABP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR