Tisk…

Anti-MIF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5571-30TEA 2510 CZK
BIOV5571-30T
Anti-MIF Rabbit Polyclonal Antibody
Protilátky
MIF (macrophage migration inhibitory factor) was one of the first cytokine activities to be discovered and was initially described as a T cell-derived factor that inhibits the random migration of microphages. Recently, MIF was rediscovered as a pituitary hormone that acts as the counter-regulatory hormone for glucocorticoid action within the immune system. MIF was released from macrophages and T-cells in response to physiological concentrations of glucocorticoids. The secreted MIF counter-regulates the immunosuppressive effects of steroids on immune cell activation and cytokine production. MIF also plays a critical role in the host control of inflammation and immunity.

Type: Primary
Antigen: MIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR