Tisk…

Anti-RETNLB Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5654-30TEA 2510 CZK
BIOV5654-30T
Anti-RETNLB Rabbit Polyclonal Antibody
Protilátky
Human RELM beta (Resistin-like molecule beta/FIZZ2) is a new member to the family of adipocyte secreted proteins called adipocytokines. This family includes the RELM alpha, RELM beta and Resistin molecules. Interestingly, RELM beta and Resistin share similar characteristics such as an additional cysteine residue within the variable N-terminal region and are both homodimeric proteins. However, the RELM beta is expressed only in the gastrointestinal track; especially the colon, while the Resistin and RELM beta are secreted exclusively by adipocytes. Currently, the biological function of these proteins, as well as their molecular targets is largely unknown. Recombinant Human RELM beta is a disulfide-linked homodimer with a total molecular weight of 11.0 kDa, consisting of 90 amino acid residue chains.

Type: Primary
Antigen: RETNLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR