Tisk…

Anti-CBX5 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6860-25EA 7760 CZK
BIOV6860-25
Anti-CBX5 Rabbit Polyclonal Antibody
Protilátky
CBX5/HP1 alpha is a component of heterochromatin. It recognizes and binds to histone H3 tails methylated at ‘Lys-9’, leading to epigenetic repression of transcription. HP1 alpha may also interact with lamin B receptor (LBR), thereby contributing to the association of the heterochromatin with the inner nuclear membrane. Further, HP1 alpha is involved in the formation of a functional kinetochore through interaction with MIS12 complex proteins.

Type: Primary
Antigen: CBX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR