Tisk…

Anti-IgG Rabbit Antibody (HRP (Horseradish Peroxidase))

Dodavatel: BioVision
BIOV6912-100EA 5500 CZK
BIOV6912-100
Anti-IgG Rabbit Antibody (HRP (Horseradish Peroxidase))
Protilátky
Rabbit Polyclonal Secondary Antibody to Sheep IgG (H&L) HRP.

Type: Secondary
Antigen: IgG
Clonality: Secondary antibody
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: Sheep IgG
Reactivity: Sheep
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR