Tisk…

Recombinant IgG4, kappa Isotype Control [clone: MOPC-22]

Dodavatel: BioVision
BIOVA1101-200EA 18890 CZK
BIOVA1101-200
Recombinant IgG4, kappa Isotype Control [clone: MOPC-22]
Protilátky
Recombinant monoclonal antibody to Unknown Specificity (Isotype control). Manufactured using Recombinant technology with variable regions (i.e. specificity) from the hybridoma MOPC-21. This is a chimeric antibody created as part of a panel offering antibodies of the same specificity in different formats (species, isotype, subtype and modified versions) for use as isotype controls.

Type: Control
Antigen: IgG4
Clonality: Monoclonal
Clone: MOPC-22
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG4, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR