Tisk…

Anti-LYZ Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOVA1110-100EA 8430 CZK
BIOVA1110-100
Anti-LYZ Rabbit Polyclonal Antibody
Lysozymes have primarily a bacteriolytic function; those in tissues and body fluids are associated with the monocyte-macrophage system and enhance the activity of immunoagents. It has an identical amino acid sequence and physic-chemical properties of native lysozyme. The enzyme cleaves b (1,4) linkage between N-acetylglucosamine and N-acetylmuramic acid of the peptidoglycan component in the bacterial cell wall. It exhibits four times more activity than commercial chicken lysozyme and is useful in the recovery of proteins expressed in bacteria (e.g. E. coli). EZ Lysozyme is designed for gentle cell lysis which leads to a minimal risk of protein denaturation.

Type: Primary
Antigen: LYZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR