Tisk…

Anti-VEGFD Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5643-30TEA 2850 CZK
BIOV5643-30T
Anti-VEGFD Rabbit Polyclonal Antibody
EGF-D is most closely related to VEGF-C by virtue of the presence of N- and C-terminal extensions that are not found in other VEGF family members. In adult human tissues, VEGF-D mRNA is most abundant in heart, lung, skeletal muscle, colon, and small intestine. Analyses of VEGF-D receptor specificity revealed that VEGF-D is a ligand for both VEGF receptors (VEGFRs) VEGFR-2 (Flk1) and VEGFR-3 (Flt4) and can activate these receptors. However, VEGF-D does not bind to VEGFR-1. Expression of a truncated derivative of VEGF-D demonstrated that the receptor-binding capacities reside in the portion of the molecule that is most closely related in primary structure to other VEGF family members and that corresponds to the mature form of VEGF-C. In addition, VEGF-D is a mitogen for endothelial cells. The recombinant VEGF-D as the immunogen was expressed based on sequence from Genebank Accession# D89630, 20-354 aa.

Type: Primary
Antigen: VEGFD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR