Tisk…

Anti-CD16/CD32 Recombinant Monoclonal Antibody [clone: 2.4G2]

Dodavatel: BioVision
BIOVA1107-200EA 19880 CZK
BIOVA1107-200
Anti-CD16/CD32 Recombinant Monoclonal Antibody [clone: 2.4G2]
Recombinant monoclonal antibody to CD16/CD32. Manufactured using recombinant technology with variable regions (i.e. specificity) from the hybridoma 2.4G2. This antibody binds murine Fc Receptors CD16/CD32 and inhibits binding of antibodies to these. It can therefore be used as an Fc-blocking reagent, to reduce non-specific staining of cells by antibodies binding via their Fc-region.

Type: Primary
Antigen: CD16/CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG, kappa
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR