Order Entry
Anti-CD68 Mouse Monoclonal Antibody (CF488A) [clone: LAMP4/824]
Anti-CD68 Mouse Monoclonal Antibody (CF488A) [clone: LAMP4/824]
 :  Biotium
Anti-CD68 Mouse Monoclonal Antibody (CF488A) [clone: LAMP4/824]
 :  Biotium
  • Catalog No:
  • BNC880824-100
  • BNC880824-500
  • Antigen symbol:
  • CD68
  • CD68
  • Antigen synonyms:
  • Scavenger Receptor Class D Member-1|Microsialin|GP110|SCARD1|LAMP4|Macrosialin
  • Scavenger Receptor Class D Member-1|Microsialin|GP110|SCARD1|LAMP4|Macrosialin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone LAMP4/824
  • Clone LAMP4/824
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 647419
  • 647419
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide
  •  
    Size
    Pk
  • BNC880824-100
    BNC880824-100
    100 μL
    100 µl
    1 * 100 µl
    1 * 100 µl - 5 770,00 Kč
    5 770,00 Kč
    1 * 100 µl
       5 770,00 Kč
     : BNC880824-100
     : BTIUBNC880824-100
    Specifications
    Antigen symbol
    CD68
    Environmentally Preferable
    Antigen synonyms
    Scavenger Receptor Class D Member-1|Microsialin|GP110|SCARD1|LAMP4|Macrosialin
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone LAMP4/824
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    647419
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide
  • BNC880824-500
    BNC880824-500
    500 μL
    500 µl
    1 * 500 µl
    1 * 500 µl - 15 940,00 Kč
    15 940,00 Kč
    1 * 500 µl
       15 940,00 Kč
     : BNC880824-500
     : BTIUBNC880824-500
    Specifications
    Antigen symbol
    CD68
    Environmentally Preferable
    Antigen synonyms
    Scavenger Receptor Class D Member-1|Microsialin|GP110|SCARD1|LAMP4|Macrosialin
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 488A
    Clone
    Clone LAMP4/824
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    647419
    Isotype
    IgG1 kappa
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0,1 mg/ml
    Storage buffer
    PBS/0,1% BSA/0,05% azide