VWR®, Accessories for Standard and Advanced Shakers
Supplier: VWR Collection
Certificates
About this item
Specifications
- Catalog No:
- 444-2948
- 444-2947
- 444-2950
- 444-2949
- 444-2942
- 444-2941
- 444-2930
- 444-2946
- 444-2945
- 444-2931
- 444-2940
- 444-2936
- 444-2935
- 444-2937
- 444-2932
- 444-2934
- 444-2933
- 444-2970
- 444-2953
- 444-2943
- 444-2944
- 444-7087
- 444-2952
- 444-2951
- Platform W×D:
- 457×457 mm
- 914×610 mm
- 610×457 mm
- 610×610 mm
- 330×279 mm
Specifications
Product Information
- Catalog #ForPkAvailabilityPrice
- Large vessel carrier platform, has 4 adjustable cushioned rollers to hold glassware securely, 764×459×353 mm, bar size 457 mmLarge vessel carrier platform, has 4 adjustable cushioned rollers to hold glassware securely, 764×459×353 mm, bar size 457 mm444-2940Standard/Advanced 5000 shakers11 * 1 KS43 590,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Large vessel carrier platform, has 4 adjustable cushioned rollers to hold glassware securely, 619×617×360 mm, bar size 610 mmLarge vessel carrier platform, has 4 adjustable cushioned rollers to hold glassware securely, 619×617×360 mm, bar size 610 mm444-2941Advanced 10000-1/10000-211 * 1 KS45 530,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Large vessel carrier platform, has 5 adjustable cushioned rollers to hold glassware securely, 937×617×360 mm, bar size 610 mmLarge vessel carrier platform, has 5 adjustable cushioned rollers to hold glassware securely, 937×617×360 mm, bar size 610 mm444-2942Advanced 15000-1/15000-211 * 1 KS59 320,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Replacement 457 mm roller bar with mounting hardwareReplacement 457 mm roller bar with mounting hardware444-2943Use with carrier 444-2940 or platforms 444-2938 / 444-293911 * 1 KS1 940,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Replacement 610 mm roller bar with mounting hardwareReplacement 610 mm roller bar with mounting hardware444-2944Use with carriers 444-2941 / 444-294211 * 1 KS2 430,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Universal platform, 330×330 mmUniversal platform, 330×330 mm444-2930Standard/Advanced 3500, Advanced 3750 shakers11 * 1 KS7 120,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Universal platform, 457×457 mm*Universal platform, 457×457 mm*444-2931Standard/Advanced 3500, Advanced 3750** shakers11 * 1 KS11 790,00 Kč
Specifications
Environmentally PreferablePlatform W×D457×457 mm - Universal platform 330×279 mmUniversal platform 330×279 mm444-7087Standard/Advanced 3500, Incubating 3500, Advanced 3750, Professional 3500 shakers, mounting Erlenmeyer flask clamps and test tube racks11 * 1 KS8 500,00 Kč
Specifications
Environmentally PreferablePlatform W×D330×279 mm - Universal platform, 610×457 mm*Universal platform, 610×457 mm*444-2932Standard/Advanced 3500, Advanced 3750**, Standard/Advanced 5000** shakers11 * 1 KS12 360,00 Kč
Specifications
Environmentally PreferablePlatform W×D610×457 mm - Universal platform, 610×610 mmUniversal platform, 610×610 mm444-2933Advanced 10000-1/10000-211 * 1 KS30 080,00 Kč
Specifications
Environmentally PreferablePlatform W×D610×610 mm - Universal platform, 762×457 mm*Universal platform, 762×457 mm*444-2934Standard/Advanced 5000 shakers11 * 1 KS28 970,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Culture platform, 457×457 mm*Culture platform, 457×457 mm*444-2936Standard/Advanced 3500, Advanced 3750** shakers11 * 1 KS10 820,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Universal platform, 914×610 mmUniversal platform, 914×610 mm444-2935Advanced 15000-1/15000-211 * 1 KS39 460,00 Kč
Specifications
Environmentally PreferablePlatform W×D914×610 mm - Culture platform, 610×457 mm*Culture platform, 610×457 mm*444-2937Standard/Advanced 3500, Advanced 3750**, Standard/Advanced 5000 shakers11 * 1 KS14 850,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 16×125 ml flask clamps, 330×330 mmDedicated platform with 16×125 ml flask clamps, 330×330 mm444-2945Standard/Advanced 3500, Advanced 3750 shakers11 * 1 KS19 340,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 12×250 ml flask clamps, 330×330 mmDedicated platform with 12×250 ml flask clamps, 330×330 mm444-2946Standard/Advanced 3500, Advanced 3750 shakers11 * 1 KS19 340,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 8×500 ml flask clamps, 330×330 mmDedicated platform with 8×500 ml flask clamps, 330×330 mm444-2947Standard/Advanced 3500, Advanced 3750 shakers11 * 1 KS19 340,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 4×1 L flask clamps, 330×330 mmDedicated platform with 4×1 L flask clamps, 330×330 mm444-2948Standard/Advanced 3500, Advanced 3750 shakers11 * 1 KS19 340,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 27×125 ml flask clamps, 457×457 mm*Dedicated platform with 27×125 ml flask clamps, 457×457 mm*444-2949Standard/Advanced 3500, Advanced 3750** shakers11 * 1 KS23 900,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 20×250 ml flask clamps, 457×457 mm*Dedicated platform with 20×250 ml flask clamps, 457×457 mm*444-2950Standard/Advanced 3500, Advanced 3750** shakers11 * 1 KS23 180,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 13×500 ml flask clamps, 457×457 mm*Dedicated platform with 13×500 ml flask clamps, 457×457 mm*444-2951Standard/Advanced 3500, Advanced 3750** shakers11 * 1 KS21 440,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Dedicated platform with 9×1 L flask clamps, 457×457 mmDedicated platform with 9×1 L flask clamps, 457×457 mm444-2952Standard/Advanced 3500, Advanced 3750** shakers11 * 1 KS21 440,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Platform for separating funnels, 457×457 mmPlatform for separating funnels, 457×457 mm444-2953Standard/Advanced 3500, Advanced 3750 shakers11 * 1 KS20 890,00 Kč
Specifications
Environmentally PreferablePlatform W×D - Clamp for separating funnelsClamp for separating funnels444-2970Use with platforms 444-2931/444-295311 * 1 KS6 160,00 Kč
Specifications
Environmentally PreferablePlatform W×D