Tisk…
Handheld refractometers, OPTi
Dodavatel: Bellingham + Stanley
635-0622EA 0 CZK
BELL38-83
Handheld refractometers, OPTi
The OPTi are quality instruments designed for a wide range of industrial applications.
  • Shallow prism dish made of 316 stainless steel
  • Zero calibration with distilled water
  • Automated temperature compensation (ATC)
Informace o dodávce: Supplied with two batteries, multi-language guide, certificate of calibration and protective case.