Tisk…
Labels for LABXPERT™ and IDXPERT™ printers, type B-461
Dodavatel: Brady
BRDY060942EA 2790 CZK
BRDY060942
Labels for LABXPERT™ and IDXPERT™ printers, type B-461
Printable, self-laminating polyester labels for LABXPERT™ and IDXPERT™ printers. Certain auto formatting features may not be enabled when using this part in an IDXPERT™ printer.
  • Printable high-performance laboratory self-laminating polyester
  • Ultra-thin construction prevents jamming in racks and centrifuges
  • Suitable for liquid nitrogen application
Ideal for labelling cryo, centrifuge and PCR tubes that will be exposed to extreme temperatures in the laboratory and holds-up against common label removal chemicals and sized to fit the most popular tube sizes.