Tisk…
Labels for BBP33 printers, type B-425, WorkHorse™
Dodavatel: Brady
WorkHorse
BRDY142892EA 4220 CZK
BRDY142892 BRDY142891 BRDY361733 BRDY361734 BRDY143053 BRDY142962
Labels for BBP33 printers, type B-425, WorkHorse™
Štítky
These polypropylene labels have a matt finish and are used for product and general purpose identification, general labeling for indoor use that requires outstanding chemical and solvent resistance.