Tisk…
Anti-HPV16L1 Mouse Monoclonal Antibody [clone: HPV16L1/1058]
Dodavatel: Abnova
ABNOMAB14430EA 14770 CZK
ABNOMAB14430
Anti-HPV16L1 Mouse Monoclonal Antibody [clone: HPV16L1/1058]
Protilátky
Mouse monoclonal antibody raised against human papilloma virus 16 (HPV-16).
Type: Primary
Antigen: HPV16L1
Clonality: Monoclonal
Clone: HPV16L1/1058
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity:
Objednat