Tisk…

Anti-CNN1 Monoclonal Antibody [clone: rCNN1/832]

Dodavatel: Abnova
ABNORAB00298EA 14030 CZK
ABNORAB00298
Anti-CNN1 Monoclonal Antibody [clone: rCNN1/832]
Protilátky
Recombinant mouse monoclonal antibody raised against human CNN1.

Type: Primary
Antigen: CNN1
Clonality: Monoclonal
Clone: rCNN1/832
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR