Tisk…

Anti-CAPN1 Rabbit Polyclonal Antibody

Dodavatel: Abnova
ABNOPAB30753EA 15540 CZK
ABNOPAB30753
Anti-CAPN1 Rabbit Polyclonal Antibody
Protilátky
Rabbit polyclonal antibody raised against partial recombinant human CAPN1.

Type: Primary
Antigen: CAPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR