Tisk…

Anti-CHGA Monoclonal Antibody [clone: CHGA/1773R]

Dodavatel: Abnova
ABNORAB00321EA 14030 CZK
ABNORAB00321
Anti-CHGA Monoclonal Antibody [clone: CHGA/1773R]
Protilátky
Recombinant rabbit monoclonal antibody raised against human CHGA.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: CHGA/1773R
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR