Tisk…

Anti-GYPA Monoclonal Antibody [clone: GYPA/1725R]

Dodavatel: Abnova
ABNORAB00329EA 14030 CZK
ABNORAB00329
Anti-GYPA Monoclonal Antibody [clone: GYPA/1725R]
Protilátky
Recombinant rabbit monoclonal antibody raised against human GYPA.

Type: Primary
Antigen: GYPA
Clonality: Monoclonal
Clone: GYPA/1725R
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR