Tisk…

Anti-IFNG Mouse Monoclonal Antibody [clone: IFNG/466]

Dodavatel: Abnova
ABNOMAB14562EA 14770 CZK
ABNOMAB14562
Anti-IFNG Mouse Monoclonal Antibody [clone: IFNG/466]
Protilátky
Mouse monoclonal antibody raised against full length recombinant human IFNG.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: IFNG/466
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR