Tisk…
BIOV1649
BIOV1649-1GEA 1910 CZK
BIOV1649-1G BIOV1649-50G BIOV1649-5G
BIOV1649
Induces β-galactosidase activity in many bacteria. Functioning as a lac analog, IPTG induces β -galactosidase activity by binding to and inhibiting the lac repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ.