Accessories
Dodavatel: Thermo Fisher Scientific
ORIO1010005EA 3930 CZK
ORIO1010005
Accessories
Meřiče elektrochemické Meřiče pH a multimetry
This cable adapter can be used to connect a USB port to the RS232 port of a meter.
The adapter allows interface between a USB connection and Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters and legacy Orion™ Star and Star Plus meters.