Tisk…

ASB-14 ≥97.0%

Dodavatel: BioVision

BIOV1738-5GEA 4940 CZK
BIOV1738-5G BIOV1738-25G
ASB-14 ≥97.0%
ASB-14

A zwitterionic detergent useful for the solubilisation of proteins including previously undetected membrane proteins. ASB-14 exhibits better protein solubilisation properties than CHAPS. Also useful for solubilising proteins for 2D-gel electrophoresis.


  • Synonyms: 3-[N,N-Dimethyl(3-myristoylaminopropyl)ammonio]propanesulphonate; Amidosulfobetaine-14

Vzorec: C₂₂H₄₆N₂O₄S
CAS číslo: 216667-08-2

Objednat

Výsledky specifikačního testu

Appearance White to off-white solid
Purity ≥97% by TLC
Solubility H₂O (50 mg/ml)
Storage +4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR