Tisk…
Bovine native collagen (from Achilles tendon)
Dodavatel: Abnova
ABNOP5215EA 3770 CZK
ABNOP5215
Bovine native collagen (from Achilles tendon)
Proteiny
Collagen is an inert, rigid protein found predominantly in skin, ligaments, bones and teeth. It is a lyophilised, shredded insoluble preparation, used as a substrate for collagenase.
Its most distinctive characteristic in its role as a transmitter of mechanical force is its inelasticity. It is stable for at least three months.