Tisk…

Anti-RFC3 Rabbit Polyclonal Antibody [clone: L227]

Dodavatel: Bioworld Technology
BWRLBS3511EA 8030 CZK
BWRLBS3511
Anti-RFC3 Rabbit Polyclonal Antibody [clone: L227]
Protilátky
Synthetic peptide, corresponding to amino acids 200-250 of Human RFC3.

RFC3 (L227) pAb detects endogenous levels of RFC3 protein.

Type: Primary
Antigen: RFC3
Clonality: Polyclonal
Clone: L227
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR