Tisk…
Drums, wide neck
Dodavatel: Semadeni
SEMA81061110EA 10420 CZK
SEMA81061110 SEMA81091110
Drums, wide neck
Sudy
Robust and durable wide neck drums, made of HDPE, natural-white.
  • Thick walled
  • Temperature resistant from –30 to + 70 °C
  • Lids available on request
The barrels are used wherever hygiene is required, such as the meat industry and the entire food industry. In many cases to store and transport solid and liquid media.