Tisk…

Anti-REC8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-7726R-HRPEA 24980 CZK
BOSSBS-7726R-HRP
Anti-REC8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Required during meiosis for separation of sister chromatids and homologous chromosomes. Proteolytic cleavage of REC8 on chromosome arms by separin during anaphase I allows for homologous chromosome separation in meiosis I and cleavage of REC8 on centromeres during anaphase II allows for sister chromatid separation in meiosis II.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: REC8
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR