Tisk…

Anti-ZNF830 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BOSSBS-7782R-CY5.5EA 24190 CZK
BOSSBS-7782R-CY5.5
Anti-ZNF830 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
This gene encodes for a coiled coil domain containing 16 protein and is located on chromosome 17. The protein may act as a regulator of the cell cycle in embryos by participating in control of M phase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF830
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR