Tisk…

Anti-NEK8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-7815R-A647EA 24190 CZK
BOSSBS-7815R-A647
Anti-NEK8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Required for renal tubular integrity.Tissue specificity: Highest expression in thyroid, adrenal gland and skin. Low levels in spleen, colon and uterus. Overexpressed in breast tumors, with highest expression in infiltrating ductal carcinomas and moderate levels in mucinous adenocarcinoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NEK8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR