Tisk…

Anti-RABGAP1 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BOSSBS-7826R-CY5.5EA 24190 CZK
BOSSBS-7826R-CY5.5
Anti-RABGAP1 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
May act as a GTPase-activating protein of RAB6A. May play a role in microtubule nucleation by centrosome. May participate in a RAB6A-mediated pathway involved in the metaphase-anaphase transition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RABGAP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR