Tisk…

Anti-MFAP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-7810R-HRPEA 24190 CZK
BOSSBS-7810R-HRP
Anti-MFAP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Microfibrillar-associated protein 1; MFAP1_HUMAN.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: MFAP1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR