Tisk…

Anti-SUGT1 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BOSSBS-7831R-CY5EA 24190 CZK
BOSSBS-7831R-CY5
Anti-SUGT1 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
This gene is homologous to the yeast gene SGT1, which encodes a protein involved in kinetochore function and required for the G1/S and G2/M transitions. Complementation studies suggest that the human protein has similar functions. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SUGT1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR